Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128006.2 | NP_001121478.1 | 5 - 6 | 436 | 81 | |
GenBank mRNA | AL888583.2 | - | 5 - 6 | 386 | 81 | |
BC166252.1 | - | 5 - 6 | 389 | 81 | ||
BC168452.1 | - | 4 - 5 | 323 | 81 | ||
CU024953.1 | - | 4 - 5 | 365 | 81 | ||
CX917894.1 | - | 5 - 6 | 427 | 81 | ||
CX928579.1 | - | 5 - 6 | 436 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |