Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128009.1 | NP_001121481.1 | 1313 | 94 | ||
GenBank mRNA | BC166255.1 | - | 1313 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis |