Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam234a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128045.1 | NP_001121517.1 | 2 - 3 | 388 | 85 | |
XM_012970131.2 | XP_012825585.1 | 2 - 3 | 417 | 85 | ||
XM_012970133.2 | XP_012825587.1 | 2 - 3 | 413 | 85 | ||
XM_018097045.1 | XP_017952534.1 | 2 - 3 | 323 | 85 | ||
GenBank mRNA | BC166371.1 | - | 2 - 3 | 388 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |