Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam234a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128045.1 | NP_001121517.1 | 4 - 5 | 686 | 91 | |
XM_012970131.2 | XP_012825585.1 | 4 - 5 | 715 | 91 | ||
XM_012970133.2 | XP_012825587.1 | 4 - 5 | 711 | 91 | ||
XM_018097045.1 | XP_017952534.1 | 4 - 5 | 621 | 91 | ||
GenBank mRNA | BC166371.1 | - | 4 - 5 | 686 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |