Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pard6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128639.1 | NP_001122111.1 | 2 - 3 | 357 | 62 | |
GenBank mRNA | BC167691.1 | - | 2 - 3 | 357 | 62 | |
CR760394.2 | - | 2 - 3 | 360 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |