Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128661.1 | NP_001122133.1 | 4 - 5 | 1161 | 81 | |
XM_012967841.2 | XP_012823295.1 | 4 - 5 | 1221 | 81 | ||
XM_018096098.1 | XP_017951587.1 | 4 - 5 | 1221 | 81 | ||
GenBank mRNA | BC135908.1 | - | 4 - 5 | 1092 | 81 | |
BC167699.1 | - | 4 - 5 | 1161 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |