Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128661.1 | NP_001122133.1 | 5 - 6 | 1303 | 99 | |
XM_012967841.2 | XP_012823295.1 | 5 - 6 | 1363 | 99 | ||
XM_018096098.1 | XP_017951587.1 | 5 - 6 | 1363 | 99 | ||
GenBank mRNA | BC135908.1 | - | 5 - 6 | 1234 | 99 | |
BC167699.1 | - | 5 - 6 | 1303 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |