Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: angptl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129936.1 | NP_001123408.1 | 2 - 3 | 905 | 59 | |
GenBank mRNA | BC167403.1 | - | 2 - 3 | 905 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |