Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c7orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130201.1 | NP_001123673.1 | 4 - 5 | 451 | 128 | |
GenBank mRNA | BC166964.1 | - | 4 - 5 | 451 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |