Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130343.3 | NP_001123815.2 | 3 - 4 | 2840 | 61 | |
GenBank mRNA | DT443629.1 | - | 1 - 2 | 135 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |