Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130350.1 | NP_001123822.1 | 2 - 3 | 819 | 94 | |
XM_012958656.2 | XP_012814110.1 | 3 - 4 | 977 | 94 | ||
XM_012958657.2 | XP_012814111.1 | 3 - 4 | 1004 | 94 | ||
GenBank mRNA | BC167630.1 | - | 2 - 3 | 819 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |