Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ciao2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134804.1 | NP_001128276.1 | 3 - 4 | 366 | 64 | |
GenBank mRNA | BC089077.1 | - | 3 - 4 | 333 | 64 | |
BC168797.1 | - | 3 - 4 | 366 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |