Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142083.1 | NP_001135555.1 | 2 - 3 | 321 | 98 | |
GenBank mRNA | BC168039.1 | - | 2 - 3 | 321 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |