Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142100.2 | NP_001135572.1 | 6 - 7 | 557 | 73 | |
XM_012954789.2 | XP_012810243.1 | 5 - 6 | 1564 | 73 | ||
XM_012954790.2 | XP_012810244.1 | 5 - 6 | 1564 | 73 | ||
GenBank mRNA | BC168076.1 | - | 6 - 7 | 559 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |