Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldh5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142106.1 | NP_001135578.1 | 7 - 8 | 1313 | 97 | |
XM_018094569.1 | XP_017950058.1 | 6 - 7 | 1231 | 97 | ||
GenBank mRNA | BC124570.1 | - | 7 - 8 | 1662 | 97 | |
BC168085.1 | - | 7 - 8 | 1313 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |