Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hacd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142156.1 | NP_001135628.1 | 633 | 68 | ||
GenBank mRNA | BC168480.1 | - | 633 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |