Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142170.3 | NP_001135642.3 | 5 - 6 | 890 | 71 | |
GenBank mRNA | BC168496.1 | - | 5 - 6 | 890 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |