Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eci2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142224.1 | NP_001135696.1 | 4 - 5 | 495 | 87 | |
XM_012964399.2 | XP_012819853.1 | 5 - 6 | 668 | 87 | ||
XM_012964400.2 | XP_012819854.1 | 4 - 5 | 596 | 87 | ||
GenBank mRNA | BC168804.1 | - | 4 - 5 | 495 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |