Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iqcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142226.2 | NP_001135698.2 | 2 - 3 | 816 | 100 | |
GenBank mRNA | BC168810.1 | - | 2 - 3 | 761 | 100 | |
CR855451.2 | - | 2 - 3 | 777 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |