Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iqcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142226.2 | NP_001135698.2 | 4 - 5 | 1125 | 90 | |
GenBank mRNA | BC168810.1 | - | 4 - 5 | 1070 | 90 | |
CR855451.2 | - | 4 - 5 | 1086 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |