Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142239.2 | NP_001135711.2 | 4 - 5 | 390 | 98 | |
GenBank mRNA | BC168838.1 | - | 4 - 5 | 384 | 98 | |
BX703151.1 | - | 4 - 5 | 389 | 98 | ||
CU025080.1 | - | 4 - 5 | 390 | 98 | ||
DR879072.1 | - | 4 - 5 | 366 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |