Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142899.1 | NP_001136371.1 | 2 - 3 | 193 | 93 | |
GenBank mRNA | BC087773.1 | - | 2 - 3 | 178 | 93 | |
BC135208.1 | - | 2 - 3 | 168 | 93 | ||
BC157546.1 | - | 2 - 3 | 193 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |