Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142899.1 | NP_001136371.1 | 3 - 4 | 251 | 84 | |
GenBank mRNA | BC087773.1 | - | 3 - 4 | 236 | 84 | |
BC135208.1 | - | 3 - 4 | 226 | 84 | ||
BC157546.1 | - | 3 - 4 | 251 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |