Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142899.1 | NP_001136371.1 | 4 - 5 | 418 | 104 | |
GenBank mRNA | BC087773.1 | - | 4 - 5 | 403 | 104 | |
BC135208.1 | - | 4 - 5 | 393 | 104 | ||
BC157546.1 | - | 4 - 5 | 418 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |