Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sirt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142909.1 | NP_001136381.1 | 7 - 8 | 1706 | 111 | |
XM_012966096.2 | XP_012821550.1 | 8 - 9 | 2226 | 111 | ||
GenBank mRNA | BC161420.1 | - | 7 - 8 | 1706 | 111 | |
CR926386.2 | - | 8 - 9 | 1790 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |