Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143925.1 | NP_001137397.1 | 5 - 6 | 528 | 89 | |
XM_018092949.1 | XP_017948438.1 | 5 - 6 | 711 | 89 | ||
GenBank mRNA | CR761197.2 | - | 5 - 6 | 623 | 89 | |
EU422982.1 | - | 5 - 6 | 528 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Xenopus tropicalis Probe spans exons |