Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sptssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005001.1 | NP_001005001.1 | 2 | 447 | 71 | |
GenBank mRNA | BC076647.1 | - | 2 | 447 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |