Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b3galt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016095.3 | NP_001016095.2 | 1 | 1043 | 83 | |
GenBank mRNA | BC090565.1 | - | 1 | 1043 | 83 | |
CR762172.2 | - | 1 | 966 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |