Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aplnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032321.1 | NP_001027492.1 | 1 | 1046 | 95 | |
GenBank mRNA | BC096504.1 | - | 1 | 1046 | 95 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |