Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100281664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154584.1 | NP_001148056.1 | 1604 | 100 | ||
GenBank mRNA | EU957485.1 | - | 1604 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |