Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: saf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111963.1 | NP_001105433.1 | 124 | 63 | ||
GenBank mRNA | X58573.1 | - | 124 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Zea mays |