Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cesa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112202.1 | NP_001105672.1 | 656 | 114 | ||
GenBank mRNA | AY372244.1 | - | 656 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Zea mays |