Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112378.1 | NP_001105848.1 | 1274 | 57 | ||
GenBank mRNA | BT039346.1 | - | 1186 | 57 | ||
DQ327709.1 | - | 1274 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |