Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136824.1 | NP_001130296.1 | 812 | 70 | ||
GenBank mRNA | BT033466.1 | - | 812 | 70 | ||
BT033823.1 | - | 838 | 70 | |||
BT054961.1 | - | 812 | 70 | |||
BT066424.1 | - | 812 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Zea mays |