Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100285243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158137.1 | NP_001151609.1 | 345 | 109 | ||
GenBank mRNA | EU971391.1 | - | 345 | 109 |