Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001147546.1 | NP_001141018.1 | 717 | 104 | ||
GenBank mRNA | BT036250.1 | - | 717 | 104 | ||
BT040176.1 | - | 984 | 104 | |||
BT043225.1 | - | 807 | 104 | |||
BT064266.1 | - | 860 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Zea mays |