Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111538.1 | NP_001105008.1 | 1252 | 96 | ||
GenBank mRNA | AF290414.1 | - | 1252 | 96 |