Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111616.1 | NP_001105086.1 | 326 | 88 | ||
GenBank mRNA | AF458962.1 | - | 326 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Zea mays |