Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gsh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111672.1 | NP_001105142.1 | 1021 | 71 | ||
GenBank mRNA | AJ302783.1 | - | 1021 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |