Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gsh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111672.1 | NP_001105142.1 | 761 | 70 | ||
GenBank mRNA | AJ302783.1 | - | 761 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Zea mays |