Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156350.1 | NP_001149822.1 | 818 | 81 | ||
GenBank mRNA | BT040236.1 | - | 700 | 81 | ||
EU964791.1 | - | 818 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |