Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111881.1 | NP_001105351.1 | 856 | 75 | ||
GenBank mRNA | AY109538.1 | - | 855 | 75 | ||
S44171.1 | - | 856 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Zea mays |