Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pep4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111968.1 | NP_001105438.1 | 2323 | 73 | ||
GenBank mRNA | X61489.1 | - | 2323 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |