Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112035.2 | NP_001105505.2 | 1161 | 90 | ||
GenBank mRNA | AF039304.1 | - | 1181 | 90 | ||
BT039711.1 | - | 1161 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Zea mays |