Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112092.1 | NP_001105562.1 | 462 | 65 | ||
GenBank mRNA | BT065528.1 | - | 462 | 65 | ||
BT065599.1 | - | 495 | 65 | |||
EU959120.1 | - | 470 | 65 | |||
X55967.1 | - | 462 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |