Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112137.1 | NP_001105607.1 | 325 | 64 | ||
GenBank mRNA | AF035460.1 | - | 325 | 64 | ||
EU961071.1 | - | 339 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |