Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112596.1 | NP_001106066.1 | 8184 | 74 | ||
GenBank mRNA | DQ020098.1 | - | 8184 | 74 |