Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111406.1 | NP_001104876.1 | 617 | 88 | ||
GenBank mRNA | AF007793.1 | - | 617 | 88 | ||
AF250050.1 | - | 637 | 88 | |||
AY109345.1 | - | 637 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |