Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hir2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111501.1 | NP_001104971.1 | 397 | 69 | ||
GenBank mRNA | AF236374.1 | - | 397 | 69 | ||
BT083877.1 | - | 727 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |