Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111662.1 | NP_001105132.1 | 1572 | 74 | ||
GenBank mRNA | AJ277702.1 | - | 1572 | 74 | ||
BT042243.1 | - | 1816 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase 3' Most Zea mays |